5-(4-(1H-1,2,4-Triazol-1-yl)phenyl)-2H-tetrazole 98%

Supplier: Ambeed
Danger

A1154781-1G A1154781-250MG A1154781-5G
76674-702EA 75.33 USD
76674-702 76674-704 76674-706
5-(4-(1H-1,2,4-Triazol-1-yl)phenyl)-2H-tetrazole 98%
5-(4-(1H-1,2,4-Triazol-1-yl)phenyl)-2H-tetrazole

Precaución: for Research Use Only

Formula: C₉H₇N₇
MW: 213.1988 g/mol
MDL Number: MFCD31700794
CAS Number: 1423810-54-1
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR