1,3,3a1,4,6,7,9-Heptaazaphenalene-2,5,8-triamine 98%

Supplier: Ambeed
Warning

A1154777-100MG A1154777-1G A1154777-250MG A1154777-5G
76674-690EA 50.93 USD
76674-690 76674-692 76674-694 77620-188
1,3,3a1,4,6,7,9-Heptaazaphenalene-2,5,8-triamine 98%
1,3,3a1,4,6,7,9-Heptaazaphenalene-2,5,8-triamine

Precaución: for Research Use Only

Formula: C₆H₆N₁₀
MW: 218.1788 g/mol
MDL Number: MFCD00767872
CAS Number: 1502-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR