2',5'-Dimethyl-[1,1':4',1''-terphenyl]-4,4''-diamine 98%
Supplier: Ambeed
|
Danger
|
A1154775-100MG
A1154775-1G
A1154775-250MG
A1154775-5G
76674-682EA
14
USD
76674-682
76674-684
76674-686
76674-688
2',5'-Dimethyl-[1,1':4',1''-terphenyl]-4,4''-diamine 98%
2',5'-Dimethyl-[1,1':4',1''-terphenyl]-4,4''-diamine
Precaución: for Research Use Only
Formula:
C₂₀H₂₀N₂ |
MDL Number:
MFCD31700808 CAS Number: 152219-88-0 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...