4,4'-(5'-(4-(Pyridin-4-yl)phenyl)-[1,1':3',1''-terphenyl]-4,4''-diyl)dipyridine 98%

Supplier: Ambeed
Warning

A1154765-100MG A1154765-1G A1154765-250MG
76674-664EA 44.56 USD
76674-664 76674-666 76674-668
4,4'-(5'-(4-(Pyridin-4-yl)phenyl)-[1,1':3',1''-terphenyl]-4,4''-diyl)dipyridine 98%
4,4'-(5'-(4-(Pyridin-4-yl)phenyl)-[1,1':3',1''-terphenyl]-4,4''-diyl)dipyridine

Precaución: for Research Use Only

Formula: C₃₉H₂₇N₃
MW: 537.6518 g/mol
MDL Number: MFCD31700775
CAS Number: 170165-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR