5,5-Bis(4-carboxyphenyl)[1,1:3,1:4,1:3,1-quinquephenyl]-4,4-dicarboxylic acid 98%

Supplier: Ambeed
Warning

A1154762-100MG A1154762-1G A1154762-250MG A1154762-5G
76674-658EA 35 USD
76674-658 76674-660 76674-662 77866-040
5,5-Bis(4-carboxyphenyl)[1,1:3,1:4,1:3,1-quinquephenyl]-4,4-dicarboxylic acid 98%
5,5-Bis(4-carboxyphenyl)[1,1:3,1:4,1:3,1-quinquephenyl]-4,4-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₄₆H₃₀O₈
CAS Number: 1816997-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR