[1,1':3',1''-terphenyl]-3,3'',5,5''-tetracarbaldehyde 98%

Supplier: Ambeed
Warning

A1154756-100MG A1154756-250MG A1154756-1G
76674-650EA 42.44 USD
76674-650 76674-652 77620-186
[1,1':3',1''-terphenyl]-3,3'',5,5''-tetracarbaldehyde 98%
[1,1':3',1''-terphenyl]-3,3'',5,5''-tetracarbaldehyde

Precaución: for Research Use Only

Formula: C₂₂H₁₄O₄
MW: 342.3442 g/mol
MDL Number: MFCD00192696
CAS Number: 201734-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR