4'-(Pyridin-4-yl)-[1,1'-biphenyl]-3,5-dicarboxylic acid 98%

Supplier: Ambeed
Warning

A1154755-100MG A1154755-1G A1154755-250MG
76674-644EA 32.89 USD
76674-644 76674-646 76674-648
4'-(Pyridin-4-yl)-[1,1'-biphenyl]-3,5-dicarboxylic acid 98%
4'-(Pyridin-4-yl)-[1,1'-biphenyl]-3,5-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₃NO₄
MW: 319.3108 g/mol
MDL Number: MFCD31700777
CAS Number: 2022152-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR