5,5-Difluoro-1,3,7,9-tetramethyl-10-phenyl-5H-4l4,5l4-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinine-2,8-dicarbaldehyde 98%

Supplier: Ambeed
Warning

A1154754-100MG A1154754-1G A1154754-250MG
76674-642EA 88.06 USD
76674-642 77620-182 77620-184
5,5-Difluoro-1,3,7,9-tetramethyl-10-phenyl-5H-4l4,5l4-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinine-2,8-dicarbaldehyde 98%
5,5-Difluoro-1,3,7,9-tetramethyl-10-phenyl-5H-4l4,5l4-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinine-2,8-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₂₁H₁₉BF₂N₂O₂
MW: 380.1956 g/mol
CAS Number: 2022166-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR