2,5-Diethenyl-1,4-benzenedicarboxaldehyde 98%

Supplier: Ambeed
Warning

A1154753-1G A1154753-250MG A1154753-100MG
76674-638EA 150 USD
76674-638 76674-640 77592-254
2,5-Diethenyl-1,4-benzenedicarboxaldehyde 98%
2,5-Diethenyl-1,4-benzenedicarboxaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂
MW: 186.2066 g/mol
MDL Number: MFCD31580046
CAS Number: 2065232-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR