4,4',4'',4'''-(Pyrazine-2,3,5,6-tetrayl)tetrabenzoic acid 98%

Supplier: Ambeed
Warning

A1154751-100MG A1154751-1G A1154751-250MG
76674-632EA 32.89 USD
76674-632 76674-634 76674-636
4,4',4'',4'''-(Pyrazine-2,3,5,6-tetrayl)tetrabenzoic acid 98%
4,4',4'',4'''-(Pyrazine-2,3,5,6-tetrayl)tetrabenzoic acid

Precaución: for Research Use Only

Formula: C₃₂H₂₀N₂O₈
MW: 560.5098 g/mol
CAS Number: 2089016-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR