4,4'-(Anthracene-9,10-diyl)dibenzoic acid 98%

Supplier: Ambeed
Warning

A1154742-1G A1154742-250MG A1154742-5G
76674-616EA 107.16 USD
76674-616 76674-618 76674-620
4,4'-(Anthracene-9,10-diyl)dibenzoic acid 98%
4,4'-(Anthracene-9,10-diyl)dibenzoic acid

Precaución: for Research Use Only

Formula: C₂₈H₁₈O₄
MW: 418.4401 g/mol
MDL Number: MFCD07644571
CAS Number: 42824-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR