1,4-Benzenedicarboxaldehyde, 2,5-dichloro- 98%

Supplier: Ambeed
Warning

A1154740-1G A1154740-250MG
76674-614EA 115 USD
76674-614 77620-178
1,4-Benzenedicarboxaldehyde, 2,5-dichloro- 98%
1,4-Benzenedicarboxaldehyde, 2,5-dichloro-

Precaución: for Research Use Only

Formula: C₈H₄Cl₂O₂
CAS Number: 46052-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR