1,4-Benzenedicarboxaldehyde, 2,5-diethoxy- 98%

Supplier: Ambeed
Warning

A1154736-100MG A1154736-250MG
76674-610EA 47.75 USD
76674-610 77620-176
1,4-Benzenedicarboxaldehyde, 2,5-diethoxy- 98%
1,4-Benzenedicarboxaldehyde, 2,5-diethoxy-

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₄
MW: 222.2372 g/mol
MDL Number: MFCD22374552
CAS Number: 56766-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR