[1,1:4,1:3,1:4,1-Quinquephenyl]-4,4-dicarboxaldehyde, 5-(4-formyl[1,1-biphenyl]-4-yl)- 95%

Supplier: Ambeed
Warning

A1154729-250MG A1154729-1G
76674-606EA 29 USD
76674-606 77866-037
[1,1:4,1:3,1:4,1-Quinquephenyl]-4,4-dicarboxaldehyde, 5-(4-formyl[1,1-biphenyl]-4-yl)- 95%
[1,1:4,1:3,1:4,1-Quinquephenyl]-4,4-dicarboxaldehyde, 5-(4-formyl[1,1-biphenyl]-4-yl)-

Precaución: for Research Use Only

Formula: C₄₅H₃₀O₃
MDL Number: MFCD32641635
CAS Number: 805246-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR