4,4',4''-Phosphoryltribenzoic acid 98%

Supplier: Ambeed
Warning

A1154728-100MG A1154728-250MG A1154728-5G A1154728-1G
76674-600EA 11 USD
76674-600 76674-602 76674-604 77595-908
4,4',4''-Phosphoryltribenzoic acid 98%
4,4',4''-Phosphoryltribenzoic acid

Precaución: for Research Use Only

Formula: C₂₁H₁₅O₇P
MDL Number: MFCD12128954
CAS Number: 807-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR