[1,1-Biphenyl]-4-carboxaldehyde, 4-[bis(4-formyl[1,1-biphenyl]-4-yl)amino]- 98%

Supplier: Ambeed
Warning

A1154725-100MG A1154725-250MG A1154725-5G A1154725-1G
76674-594EA 22.28 USD
76674-594 76674-596 76674-598 77592-250
[1,1-Biphenyl]-4-carboxaldehyde, 4-[bis(4-formyl[1,1-biphenyl]-4-yl)amino]- 98%
[1,1-Biphenyl]-4-carboxaldehyde, 4-[bis(4-formyl[1,1-biphenyl]-4-yl)amino]-

Precaución: for Research Use Only

Formula: C₃₉H₂₇NO₃
MW: 557.6366 g/mol
MDL Number: MFCD31700766
CAS Number: 872689-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR