[1,1:2,1-Terphenyl]-4,4-dicarboxaldehyde, 4,5-bis(4-formylphenyl)- 98%

Supplier: Ambeed
Warning

A1154724-100MG A1154724-1G A1154724-250MG A1154724-5G
76674-588EA 8 USD
76674-588 76674-590 76674-592 77566-066
[1,1:2,1-Terphenyl]-4,4-dicarboxaldehyde, 4,5-bis(4-formylphenyl)- 98%
[1,1:2,1-Terphenyl]-4,4-dicarboxaldehyde, 4,5-bis(4-formylphenyl)-

Precaución: For research use only.

Formula: C₃₄H₂₂O₄
MDL Number: MFCD00183355
CAS Number: 883835-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR