4,4'-(Ethane-1,1-diyl)diphenol 97%

Supplier: Ambeed
Warning

A1154711-1G A1154711-25G A1154711-5G
76674-582EA 14.86 USD
76674-582 76674-584 76674-586
4,4'-(Ethane-1,1-diyl)diphenol 97%
4,4'-(Ethane-1,1-diyl)diphenol

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
MDL Number: MFCD00186869
CAS Number: 2081-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR