2-(1-Hydroxy-3-methylbutylidene)-5,5-dimethyl-1,3-cyclohexanedione 97%
Supplier: Ambeed
|
Warning
|
A1154615-1G
A1154615-250MG
A1154615-25G
A1154615-5G
76674-564EA
21.99
USD
76674-564
76674-566
76674-568
76674-570
2-(1-Hydroxy-3-methylbutylidene)-5,5-dimethyl-1,3-cyclohexanedione 97%
2-(1-Hydroxy-3-methylbutylidene)-5,5-dimethyl-1,3-cyclohexanedione
Precaución: for Research Use Only
Formula:
C₁₃H₂₀O₃ |
MDL Number:
MFCD01862890 CAS Number: 172611-72-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...