(E)-1-Ethyl-2-((1-ethylquinolin-2(1H)-ylidene)methyl)quinolin-1-ium iodide 98%

Supplier: Ambeed
Danger

A1154195-100MG A1154195-1G A1154195-250MG
76674-548EA 41 USD
76674-548 76674-550 76674-552
(E)-1-Ethyl-2-((1-ethylquinolin-2(1H)-ylidene)methyl)quinolin-1-ium iodide 98%
(E)-1-Ethyl-2-((1-ethylquinolin-2(1H)-ylidene)methyl)quinolin-1-ium iodide

Precaución: for Research Use Only

Formula: C₂₃H₂₃IN₂
MDL Number: MFCD00011971
CAS Number: 63902-24-9
UN: 2811
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR