(E)-2-((5-Nitrofuran-2-yl)methylene)hydrazinecarboxamide 98%

Supplier: Ambeed
Warning

Synonyms: (E)-Nitrofurazone

A1154128-250MG A1154128-1G A1154128-100MG
76674-546EA 85.94 USD
76674-546 77595-900 77620-168
(E)-2-((5-Nitrofuran-2-yl)methylene)hydrazinecarboxamide 98%
(E)-2-((5-Nitrofuran-2-yl)methylene)hydrazinecarboxamide

Precaución: for Research Use Only

Formula: C₆H₆N₄O₄
MW: 198.1362 g/mol
MDL Number: MFCD00003225
CAS Number: 112574-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR