1,3-Di(thiophen-2-yl)propane-1,3-dione 97%

Supplier: Ambeed
Warning

A1153945-100MG A1153945-1G A1153945-250MG
76674-532EA 50.93 USD
76674-532 76674-534 76674-536
1,3-Di(thiophen-2-yl)propane-1,3-dione 97%
1,3-Di(thiophen-2-yl)propane-1,3-dione

Precaución: for Research Use Only

Formula: C₁₁H₈O₂S₂
MW: 236.3100 g/mol
MDL Number: MFCD00797223
CAS Number: 1138-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR