4,6-Diphenylpyrimidine 97%

Supplier: Ambeed
Warning

A1153935-100MG A1153935-1G A1153935-250MG
76674-526EA 35 USD
76674-526 76674-528 76674-530
4,6-Diphenylpyrimidine 97%
4,6-Diphenylpyrimidine

Precaución: for Research Use Only

Formula: C₁₆H₁₂N₂
MDL Number: MFCD00234899
CAS Number: 3977-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR