(S)-2-Amino-4-(3-hydroxyguanidino)butanoic acid dihydrochloride 98%

Supplier: Ambeed
Warning

A1153835-100MG A1153835-1G A1153835-250MG
76674-520EA 65 USD
76674-520 76674-522 76674-524
(S)-2-Amino-4-(3-hydroxyguanidino)butanoic acid dihydrochloride 98%
(S)-2-Amino-4-(3-hydroxyguanidino)butanoic acid dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₄Cl₂N₄O₃
MW: 249.0957 g/mol
MDL Number: MFCD04036978
CAS Number: 291758-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR