Methyl 1-acetyl-3-methyl-1H-indazole-6-carboxylate 97%

Supplier: Ambeed
Warning

A1153677-100MG A1153677-1G A1153677-250MG
76674-508EA 37 USD
76674-508 76674-510 76674-512
Methyl 1-acetyl-3-methyl-1H-indazole-6-carboxylate 97%
Methyl 1-acetyl-3-methyl-1H-indazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₃
MDL Number: MFCD32069581
CAS Number: 2368870-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR