4-Oxazolecarboxylic acid, 2-[[(1,1-dimethylethoxy)carbonyl]amino]-, ethyl ester 95%

Supplier: Ambeed
Warning

A1153491-100MG A1153491-1G A1153491-250MG
76674-502EA 47.75 USD
76674-502 76674-504 76674-506
4-Oxazolecarboxylic acid, 2-[[(1,1-dimethylethoxy)carbonyl]amino]-, ethyl ester 95%
4-Oxazolecarboxylic acid, 2-[[(1,1-dimethylethoxy)carbonyl]amino]-, ethyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂O₅
MW: 256.26 g/mol
MDL Number: MFCD11975886
CAS Number: 244236-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR