1-Chloronaphthalen-2-amine 95%

Supplier: Ambeed
Warning

A1153469-1G A1153469-250MG
76674-498EA 23 USD
76674-498 76674-500
1-Chloronaphthalen-2-amine 95%
1-Chloronaphthalen-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₈ClN
MDL Number: MFCD00226972
CAS Number: 16452-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR