[8-(Glycidyloxy)-n-octyl]trimethoxysilane 97%

Supplier: Ambeed
Warning

Synonyms: Trimethoxy(8-(oxiran-2-ylmethoxy)octyl)silane

A1152960-1G A1152960-250MG A1152960-5G
76674-482EA 35.01 USD
76674-482 76674-484 76674-486
[8-(Glycidyloxy)-n-octyl]trimethoxysilane 97%
[8-(Glycidyloxy)-n-octyl]trimethoxysilane

Precaución: for Research Use Only

Formula: C₁₄H₃₀O₅Si
MW: 306.47 g/mol
MDL Number: MFCD29472534
CAS Number: 1239602-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR