1,3-Bis((S)-1-phenylethyl)-1H-imidazol-3-ium chloride 97%

Supplier: Ambeed
Warning

A1152690-100MG A1152690-1G A1152690-250MG
76674-476EA 62.6 USD
76674-476 76674-478 76674-480
1,3-Bis((S)-1-phenylethyl)-1H-imidazol-3-ium chloride 97%
1,3-Bis((S)-1-phenylethyl)-1H-imidazol-3-ium chloride

Precaución: for Research Use Only

Formula: C₁₉H₂₁ClN₂
MW: 312.8364 g/mol
CAS Number: 186354-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR