3,3'-Di(dibenzothiophen-4-yl)-1,1'-biphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 3,3'-Bis(dibenzo[b,d]thiophen-4-yl)-1,1'-biphenyl, 4,4'-[1,1'-Biphenyl]-3,3'-diyldi(dibenzothiophene)

A1152540-100MG A1152540-1G A1152540-250MG
76674-468EA 31.83 USD
76674-468 76674-470 76674-472
3,3'-Di(dibenzothiophen-4-yl)-1,1'-biphenyl 98%
3,3'-Di(dibenzothiophen-4-yl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₃₆H₂₂S₂
MW: 518.69 g/mol
MDL Number: MFCD32062886
CAS Number: 1128045-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR