3-Bromo-9-([1,1':4',1''-terphenyl]-4-yl)-9H-carbazole 98%

Supplier: Ambeed
Warning

A1152539-1G A1152539-250MG A1152539-5G
76674-464EA 48 USD
76674-464 76674-466 77783-316
3-Bromo-9-([1,1':4',1''-terphenyl]-4-yl)-9H-carbazole 98%
3-Bromo-9-([1,1':4',1''-terphenyl]-4-yl)-9H-carbazole

Precaución: for Research Use Only

Formula: C₃₀H₂₀BrN
MDL Number: MFCD31692913
CAS Number: 1141017-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR