BRD 4354 ditrifluoroacetate 98%

Supplier: Ambeed
Warning

Synonyms: BRD 4354 2TFA

A1152455-10MG A1152455-1MG A1152455-25MG A1152455-5MG
76674-454EA 40 USD
76674-454 76674-456 76674-458 76674-460
BRD 4354 ditrifluoroacetate 98%
BRD 4354 ditrifluoroacetate

Precaución: for Research Use Only

Formula: C₂₅H₂₅ClF₆N₄O₅
MW: 610.93 g/mol
MDL Number: MFCD32067880
CAS Number: 2444837-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR