((1,3,5-Triazine-2,4,6-triyl)tris(benzene-3,1-diyl))tris(diphenylphosphine oxide) 97%

Supplier: Ambeed
Warning

A1152387-100MG A1152387-250MG A1152387-1G
76674-450EA 27 USD
76674-450 76674-452 77866-009
((1,3,5-Triazine-2,4,6-triyl)tris(benzene-3,1-diyl))tris(diphenylphosphine oxide) 97%
((1,3,5-Triazine-2,4,6-triyl)tris(benzene-3,1-diyl))tris(diphenylphosphine oxide)

Precaución: for Research Use Only

Formula: C₅₇H₄₂N₃O₃P₃
MDL Number: MFCD32197458
CAS Number: 1646906-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR