Formamidine hydrobromide 99% (by titrimetric analysis)

Supplier: Ambeed
Warning

A1152361-1G A1152361-250MG A1152361-5G
76674-444EA 50 USD
76674-444 76674-446 76674-448
Formamidine hydrobromide 99% (by titrimetric analysis)
Formamidine hydrobromide

Precaución: for Research Use Only

Formula: CH₄N₂·HBr
MDL Number: MFCD28369274
CAS Number: 146958-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR