(2S,3R)-2-(((S)-tert-Butylsulfinyl)amino)-3-methylpent-4-enoic acid 98%

Supplier: Ambeed
Warning

A1152327-1G A1152327-25G A1152327-5G
76674-438EA 21.22 USD
76674-438 76674-440 76674-442
(2S,3R)-2-(((S)-tert-Butylsulfinyl)amino)-3-methylpent-4-enoic acid 98%
(2S,3R)-2-(((S)-tert-Butylsulfinyl)amino)-3-methylpent-4-enoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃S
MW: 233.3278 g/mol
CAS Number: 1461641-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR