(4-(6-Methyl-1,2,4,5-tetrazin-3-yl)phenyl)methanamine hydrochloride 97%

Supplier: Ambeed
Danger

Synonyms: Methyltetrazine-amine hydrochloride

A1152138-1G A1152138-100MG A1152138-250MG
76674-430EA 660.95 USD
76674-430 77570-896 77570-898
(4-(6-Methyl-1,2,4,5-tetrazin-3-yl)phenyl)methanamine hydrochloride 97%
(4-(6-Methyl-1,2,4,5-tetrazin-3-yl)phenyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClN₅
MW: 237.6888 g/mol
MDL Number: MFCD32061909
CAS Number: 1596117-29-1
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR