N-t-BOC-(R)-2-Methyl-4-Pyrrolidinone 98%

Supplier: Ambeed
Warning

Synonyms: (R)-tert-Butyl 2-methyl-4-oxopyrrolidine-1-carboxylate

A1151846-100MG A1151846-1G A1151846-250MG
76674-414EA 90.18 USD
76674-414 76674-416 76674-418
N-t-BOC-(R)-2-Methyl-4-Pyrrolidinone 98%
N-t-BOC-(R)-2-Methyl-4-Pyrrolidinone

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₃
MW: 199.247 g/mol
MDL Number: MFCD22665783
CAS Number: 1027775-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR