4'-Bromo-1,1':3',1''-terphenyl 97%

Supplier: Ambeed
Warning

A1151688-1G A1151688-250MG
76674-396EA 42.44 USD
76674-396 76674-398
4'-Bromo-1,1':3',1''-terphenyl 97%
4'-Bromo-1,1':3',1''-terphenyl

Precaución: for Research Use Only

Formula: C₁₈H₁₃Br
MW: 309.1998 g/mol
MDL Number: MFCD26401783
CAS Number: 60631-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR