Heptylphosphonic acid 98%

Supplier: Ambeed
Danger

A1151625-1G A1151625-100MG A1151625-250MG A1151625-5G
76674-386EA 122 USD
76674-386 77865-987 77865-988 77865-989
Heptylphosphonic acid 98%
Heptylphosphonic acid

Precaución: for Research Use Only

Formula: C₇H₁₇O₃P
MDL Number: MFCD00015840
CAS Number: 4721-16-8
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR