N-Methyl-N-((3R,4R)-4-methylpiperidin-3-yl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine dihydrochloride 97%

Supplier: Ambeed
Warning

A1151445-1G A1151445-250MG A1151445-25G A1151445-5G
76674-360EA 41 USD
76674-360 76674-362 76674-364 76674-366
N-Methyl-N-((3R,4R)-4-methylpiperidin-3-yl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine dihydrochloride 97%
N-Methyl-N-((3R,4R)-4-methylpiperidin-3-yl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₂₁Cl₂N₅
MDL Number: MFCD30502880
CAS Number: 1655430-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR