2-Chloro-N-ethylethanamine hydrochloride 97%

Supplier: Ambeed
Warning

A1151192-1G A1151192-250MG
76674-352EA 155 USD
76674-352 76674-354
2-Chloro-N-ethylethanamine hydrochloride 97%
2-Chloro-N-ethylethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₁Cl₂N
MDL Number: MFCD00050518
CAS Number: 4535-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR