Tris (2,2'-bipyrazine) ruthenium dichloride 98%

Supplier: Ambeed
Warning

A1150980-100MG A1150980-1G A1150980-250MG A1150980-50MG
76674-340EA 94 USD
76674-340 76674-342 76674-344 76674-346
Tris (2,2'-bipyrazine) ruthenium dichloride 98%
Tris (2,2'-bipyrazine) ruthenium dichloride

Precaución: for Research Use Only

Formula: C₂₄H₁₈Cl₂N₁₂Ru
MW: 646.4561 g/mol
CAS Number: 80925-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR