(S)-2-Amino-4-(bis(2-((3-methylbenzyl)oxy)benzyl)amino)butanoic acid hydrochloride 99%

Supplier: Ambeed
Warning

Synonyms: V-9302 HCL

A1150931-10MG A1150931-1MG A1150931-25MG A1150931-50MG
76674-332EA 49.87 USD
76674-332 76674-334 76674-336 76674-338
(S)-2-Amino-4-(bis(2-((3-methylbenzyl)oxy)benzyl)amino)butanoic acid hydrochloride 99%
(S)-2-Amino-4-(bis(2-((3-methylbenzyl)oxy)benzyl)amino)butanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₃₄H₃₉ClN₂O₄
MW: 575.1375 g/mol
MDL Number: MFCD32633537
CAS Number: 2416138-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR