2,2'-(Pyridin-2-ylmethylene)diphenol 95%

Supplier: Ambeed
Warning

A1150759-100MG A1150759-1G A1150759-250MG
76674-326EA 90.18 USD
76674-326 76674-328 76674-330
2,2'-(Pyridin-2-ylmethylene)diphenol 95%
2,2'-(Pyridin-2-ylmethylene)diphenol

Precaución: for Research Use Only

Formula: C₁₈H₁₅NO₂
MW: 277.3172 g/mol
CAS Number: 16985-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR