1,2-Dimethyl-3-propyl-1H-imidazol-3-ium hexafluorophosphate(V) 97%

Supplier: Ambeed
Warning

A1150608-25G A1150608-5G
76674-306EA 14 USD
76674-306 76674-308
1,2-Dimethyl-3-propyl-1H-imidazol-3-ium hexafluorophosphate(V) 97%
1,2-Dimethyl-3-propyl-1H-imidazol-3-ium hexafluorophosphate(V)

Precaución: for Research Use Only

Formula: C₈H₁₅F₆N₂P
MW: 284.1823 g/mol
MDL Number: MFCD28015746
CAS Number: 157310-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR