12H-Benzo[4,5]thieno[2,3-a]carbazole 99%

Supplier: Ambeed
Warning

A1150534-1G A1150534-250MG A1150534-5G
76674-288EA 47.75 USD
76674-288 76674-290 76674-292
12H-Benzo[4,5]thieno[2,3-a]carbazole 99%
12H-Benzo[4,5]thieno[2,3-a]carbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₁NS
MW: 273.3516 g/mol
MDL Number: MFCD30291519
CAS Number: 222-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR