4-(3-Bromophenyl)-6-phenyldibenzo[b,d]furan 97%

Supplier: Ambeed
Warning

A1150480-1G A1150480-5G
76674-284EA 35.01 USD
76674-284 76674-286
4-(3-Bromophenyl)-6-phenyldibenzo[b,d]furan 97%
4-(3-Bromophenyl)-6-phenyldibenzo[b,d]furan

Precaución: for Research Use Only

Formula: C₂₄H₁₅BrO
MW: 399.2793 g/mol
MDL Number: MFCD32670066
CAS Number: 2088537-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR