4-([1,1'-Biphenyl]-4-yl)-2-chloroquinazoline 97%

Supplier: Ambeed
Warning

A1150388-1G A1150388-5G
76674-264EA 21.22 USD
76674-264 76674-266
4-([1,1'-Biphenyl]-4-yl)-2-chloroquinazoline 97%
4-([1,1'-Biphenyl]-4-yl)-2-chloroquinazoline

Precaución: for Research Use Only

Formula: C₂₀H₁₃ClN₂
MW: 316.7836 g/mol
MDL Number: MFCD28967157
CAS Number: 1262866-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR