1,2-Dimethyl-3-octyl-1H-imidazol-3-ium chloride 98%

Supplier: Ambeed
Warning

A1150346-25G A1150346-5G
76674-258EA 43.5 USD
76674-258 76674-260
1,2-Dimethyl-3-octyl-1H-imidazol-3-ium chloride 98%
1,2-Dimethyl-3-octyl-1H-imidazol-3-ium chloride

Precaución: for Research Use Only

Formula: C₁₃H₂₅ClN₂
MW: 244.8040 g/mol
MDL Number: MFCD19440950
CAS Number: 1007398-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR