1,2-Dimethyl-3-octyl-1H-imidazol-3-ium bromide 98%

Supplier: Ambeed
Warning

A1150344-25G A1150344-5G
76674-254EA 40.32 USD
76674-254 76674-256
1,2-Dimethyl-3-octyl-1H-imidazol-3-ium bromide 98%
1,2-Dimethyl-3-octyl-1H-imidazol-3-ium bromide

Precaución: for Research Use Only

Formula: C₁₃H₂₅BrN₂
MW: 289.2550 g/mol
MDL Number: MFCD19440741
CAS Number: 61546-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR